Assay Details
| Gene Symbol: Cda | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500851.2 | XP_003500899.1 | 3 - 4 | 326 | 66 | |
| GenBank mRNA | JI865905.1 | - | 3 - 4 | 377 | 66 | |
More Information
Set Membership: |
|

Cda
Genomic Map