Assay Details
| Gene Symbol: Mtfmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507150.2 | XP_003507198.1 | 8 - 9 | 1004 | 61 | |
| GenBank mRNA | JI879997.1 | - | 6 - 7 | 558 | 61 | |
| JP039155.1 | - | 6 - 7 | 558 | 61 | ||
More Information
Set Membership: |
|

Mtfmt
Genomic Map