Assay Details
| Gene Symbol: Daxx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510829.2 | XP_003510877.1 | 5 - 6 | 1939 | 61 | |
| XM_007651412.1 | XP_007649602.1 | 5 - 6 | 1934 | 61 | ||
| GenBank mRNA | JI864077.1 | - | 2 - 3 | 508 | 61 | |
| JP060249.1 | - | 2 - 3 | 508 | 61 | ||
More Information
Set Membership: |
|

Daxx
Genomic Map