Assay Details
Gene Symbol: Wdr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003512814.2 | XP_003512862.1 | 10 - 11 | 1009 | 58 | |
GenBank mRNA | JI863885.1 | - | 10 - 11 | 859 | 58 | |
JP053088.1 | - | 10 - 11 | 859 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
