Assay Details
| Gene Symbol: Acadm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650206.1 | XP_007648396.1 | 4 - 5 | 439 | 65 | |
| GenBank mRNA | JP061397.1 | - | 4 - 5 | 439 | 65 | |
More Information
Set Membership: |
|

Acadm
Genomic Map