Assay Details
| Gene Symbol: Sparc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007640142.1 | XP_007638332.1 | 5 - 6 | 488 | 57 | |
| XM_007640143.1 | XP_007638333.1 | 5 - 6 | 485 | 57 | ||
| GenBank mRNA | JI872780.1 | - | 5 - 6 | 470 | 57 | |
More Information
Set Membership: |
|

Sparc
Genomic Map