Assay Details
| Gene Symbol: Trim25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007624112.1 | XP_007622302.1 | 7 - 8 | 1357 | 70 | |
| XM_007624115.1 | XP_007622305.1 | 6 - 7 | 1345 | 70 | ||
More Information
Set Membership: |
|

Trim25
Genomic Map