Assay Details
| Gene Symbol: Lphn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007640357.1 | XP_007638547.1 | 10 - 11 | 2223 | 59 | |
| XM_007640358.1 | XP_007638548.1 | 10 - 11 | 2224 | 59 | ||
| XM_007640360.1 | XP_007638550.1 | 9 - 10 | 2184 | 59 | ||
| XM_007640361.1 | XP_007638551.1 | 10 - 11 | 2026 | 59 | ||
| XM_007640362.1 | XP_007638552.1 | 9 - 10 | 1988 | 59 | ||
| XM_007640363.1 | XP_007638553.1 | 10 - 11 | 2222 | 59 | ||
| XM_007640364.1 | XP_007638554.1 | 9 - 10 | 2183 | 59 | ||
| GenBank mRNA | JI881377.1 | - | 9 - 10 | 2033 | 59 | |
More Information
Set Membership: |
|

Lphn2
Genomic Map