Assay Details
| Gene Symbol: Ssb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645424.1 | XP_007643614.1 | 2 - 3 | 223 | 65 | |
| GenBank mRNA | JI894632.1 | - | 3 - 4 | 65 | ||
| JP054552.1 | - | 2 - 3 | 214 | 65 | ||
More Information
Set Membership: |
|

Ssb
Genomic Map