Assay Details
| Gene Symbol: Lims1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007644316.1 | XP_007642506.1 | 7 - 8 | 790 | 63 | |
| GenBank mRNA | JI876909.1 | - | 7 - 8 | 885 | 63 | |
| JP061680.1 | - | 8 - 9 | 892 | 63 | ||
More Information
Set Membership: |
|

Lims1
Genomic Map