Assay Details
| Gene Symbol: Slc33a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498409.2 | XP_003498457.1 | 2 - 3 | 1235 | 68 | |
| GenBank mRNA | JI895259.1 | - | 2 - 3 | 993 | 68 | |
| JP038869.1 | - | 2 - 3 | 993 | 68 | ||
More Information
Set Membership: |
|

Slc33a1
Genomic Map