Assay Details
| Gene Symbol: Ip6k1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500343.2 | XP_003500391.1 | 3 - 4 | 786 | 60 | |
| GenBank mRNA | JI873667.1 | - | 2 - 3 | 552 | 60 | |
| JP043196.1 | - | 2 - 3 | 553 | 60 | ||
More Information
Set Membership: |
|

Ip6k1
Genomic Map