Assay Details
| Gene Symbol: Hint2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505004.2 | XP_003505052.1 | 4 - 5 | 402 | 69 | |
| GenBank mRNA | JI885428.1 | - | 1 - 2 | 75 | 69 | |
| JP057227.1 | - | 1 - 2 | 75 | 69 | ||
More Information
Set Membership: |
|

Hint2
Genomic Map