Assay Details
| Gene Symbol: Slc46a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505510.2 | XP_003505558.2 | 1 - 2 | 1115 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc46a2| Gene Symbol: Slc46a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505510.2 | XP_003505558.2 | 1 - 2 | 1115 | 75 | |
Set Membership: |
|