Assay Details
| Gene Symbol: Myd88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505920.2 | XP_003505968.1 | 3 - 4 | 1211 | 61 | |
| GenBank mRNA | JI865472.1 | - | 3 - 4 | 717 | 61 | |
| JP053884.1 | - | 3 - 4 | 717 | 61 | ||
More Information
Set Membership: |
|

Myd88
Genomic Map