Assay Details
| Gene Symbol: Rbm41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507255.2 | XP_003507303.1 | 3 - 4 | 367 | 74 | |
| XM_007647822.1 | XP_007646012.1 | 3 - 4 | 367 | 74 | ||
| XM_007647823.1 | XP_007646013.1 | 3 - 4 | 367 | 74 | ||
| GenBank mRNA | JP055784.1 | - | 3 - 4 | 378 | 74 | |
More Information
Set Membership: |
|

Rbm41
Genomic Map