Assay Details
| Gene Symbol: Hnrnpa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513851.2 | XP_003513899.1 | 8 - 9 | 1050 | 65 | |
| XM_007654064.1 | XP_007652254.1 | 9 - 10 | 1209 | 65 | ||
| GenBank mRNA | JP053583.1 | - | 1 - 2 | 155 | 65 | |
More Information
Set Membership: |
|

Hnrnpa1
Genomic Map