Assay Details
| Gene Symbol: Comt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643065.1 | XP_007641255.1 | 2 - 3 | 505 | 67 | |
| XM_007643066.1 | XP_007641256.1 | 2 - 3 | 363 | 67 | ||
| GenBank mRNA | JP055791.1 | - | 1 - 2 | 301 | 67 | |
More Information
Set Membership: |
|

Comt
Genomic Map