Assay Details
| Gene Symbol: Thg1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643427.1 | XP_007641617.1 | 3 - 4 | 500 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Thg1l| Gene Symbol: Thg1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643427.1 | XP_007641617.1 | 3 - 4 | 500 | 63 | |
Set Membership: |
|