Assay Details
| Gene Symbol: Mecp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647055.1 | XP_007645245.1 | 2 - 3 | 588 | 102 | |
| GenBank mRNA | JI870646.1 | - | 587 | 102 | ||
More Information
Set Membership: |
|

Mecp2
Genomic Map