Assay Details
| Gene Symbol: Asb17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496442.2 | XP_003496490.1 | 1 - 2 | 402 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Asb17| Gene Symbol: Asb17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496442.2 | XP_003496490.1 | 1 - 2 | 402 | 58 | |
Set Membership: |
|