Assay Details
| Gene Symbol: Gtf2f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500298.2 | XP_003500346.1 | 6 - 7 | 678 | 72 | |
| GenBank mRNA | JI883934.1 | - | 6 - 7 | 654 | 72 | |
| JP062838.1 | - | 6 - 7 | 654 | 72 | ||
More Information
Set Membership: |
|

Gtf2f2
Genomic Map