Assay Details
| Gene Symbol: Polm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504749.2 | XP_003504797.1 | 8 - 9 | 1716 | 60 | |
| XM_007645260.1 | XP_007643450.1 | 8 - 9 | 1322 | 60 | ||
| XM_007645261.1 | XP_007643451.1 | 8 - 9 | 943 | 60 | ||
| GenBank mRNA | JI891146.1 | - | 1 - 2 | 81 | 60 | |
| JP049617.1 | - | 1 - 2 | 81 | 60 | ||
More Information
Set Membership: |
|

Polm
Genomic Map