Assay Details
| Gene Symbol: Arhgap31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509149.2 | XP_003509197.1 | 3 - 4 | 620 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arhgap31| Gene Symbol: Arhgap31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509149.2 | XP_003509197.1 | 3 - 4 | 620 | 61 | |
Set Membership: |
|