Assay Details
| Gene Symbol: LOC100770113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513973.2 | XP_003514021.1 | 1 - 2 | 303 | 63 | |
| GenBank mRNA | JI862735.1 | - | 1 - 2 | 152 | 63 | |
| JP060972.1 | - | 1 - 2 | 152 | 63 | ||
More Information
Set Membership: |
|

LOC100770113
Genomic Map