Assay Details
| Gene Symbol: Ncl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653720.1 | XP_007651910.1 | 12 - 13 | 1763 | 71 | |
| XM_007653721.1 | XP_007651911.1 | 12 - 13 | 1760 | 71 | ||
| GenBank mRNA | JI878598.1 | - | 1 | 2010 | 71 | |
| JP064064.1 | - | 1 | 2010 | 71 | ||
| M15825.1 | - | 12 - 13 | 1725 | 71 | ||
More Information
Set Membership: |
|

Ncl
Genomic Map