Assay Details
| Gene Symbol: Myl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497087.2 | XP_003497135.1 | 2 - 3 | 263 | 81 | |
| XM_007607733.1 | XP_007605923.1 | 2 - 3 | 468 | 81 | ||
More Information
Set Membership: |
|

Myl9
Genomic Map