Assay Details
| Gene Symbol: Myl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497087.2 | XP_003497135.1 | 3 - 4 | 430 | 105 | |
| XM_007607733.1 | XP_007605923.1 | 3 - 4 | 635 | 105 | ||
More Information
Set Membership: |
|

Myl9
Genomic Map