Assay Details
| Gene Symbol: Stk17b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512443.2 | XP_003512491.2 | 4 - 5 | 881 | 65 | |
| GenBank mRNA | JP050512.1 | - | 4 - 5 | 881 | 65 | |
More Information
Set Membership: |
|

Stk17b
Genomic Map