Assay Details
| Gene Symbol: LOC100768961 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513005.2 | XP_003513053.2 | 2 - 3 | 476 | 84 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100768961| Gene Symbol: LOC100768961 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513005.2 | XP_003513053.2 | 2 - 3 | 476 | 84 | |