Assay Details
| Gene Symbol: Adra1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007637304.1 | XP_007635494.1 | 2 - 3 | 115 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adra1d| Gene Symbol: Adra1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007637304.1 | XP_007635494.1 | 2 - 3 | 115 | 63 | |
Set Membership: |
|