Assay Details
| Gene Symbol: P4ha3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654587.1 | XP_007652777.1 | 3 - 4 | 530 | 58 | |
| GenBank mRNA | JI893709.1 | - | 3 - 4 | 509 | 58 | |
| JP049709.1 | - | 3 - 4 | 509 | 58 | ||
More Information
Set Membership: |
|

P4ha3
Genomic Map