Assay Details
| Gene Symbol: March10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502967.2 | XP_003503015.1 | 3 - 4 | 345 | 88 | |
| XM_003502968.2 | XP_003503016.1 | 3 - 4 | 345 | 88 | ||
More Information
Set Membership: |
|

March10
Genomic Map