Assay Details
| Gene Symbol: Grb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495657.2 | XP_003495705.1 | 4 - 5 | 750 | 65 | |
| GenBank mRNA | JI870911.1 | - | 4 - 5 | 526 | 65 | |
| JP048336.1 | - | 4 - 5 | 526 | 65 | ||
More Information
Set Membership: |
|

Grb2
Genomic Map