Assay Details
| Gene Symbol: Psmg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496299.2 | XP_003496347.1 | 2 - 3 | 405 | 76 | |
| GenBank mRNA | JI870498.1 | - | 2 - 3 | 412 | 76 | |
| JP060729.1 | - | 2 - 3 | 412 | 76 | ||
More Information
Set Membership: |
|

Psmg3
Genomic Map