Assay Details
| Gene Symbol: Ppapdc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498225.2 | XP_003498273.1 | 4 - 5 | 371 | 84 | |
| GenBank mRNA | JI888476.1 | - | 3 - 4 | 199 | 84 | |
| JP064501.1 | - | 3 - 4 | 198 | 84 | ||
More Information
Set Membership: |
|

Ppapdc1b
Genomic Map