Assay Details
| Gene Symbol: Ppapdc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498225.2 | XP_003498273.1 | 6 - 7 | 662 | 77 | |
| GenBank mRNA | JI888475.1 | - | 1 - 2 | 167 | 77 | |
| JP064500.1 | - | 1 - 2 | 166 | 77 | ||
More Information
Set Membership: |
|

Ppapdc1b
Genomic Map