Assay Details
| Gene Symbol: Pigh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498267.2 | XP_003498315.1 | 1 - 2 | 200 | 55 | |
| GenBank mRNA | JP042451.1 | - | 1 - 2 | 44 | 55 | |
More Information
Set Membership: |
|

Pigh
Genomic Map