Assay Details
| Gene Symbol: Clp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506153.2 | XP_003506201.1 | 2 - 3 | 734 | 93 | |
| GenBank mRNA | JI894123.1 | - | 1 - 2 | 241 | 93 | |
| JP038722.1 | - | 1 - 2 | 241 | 93 | ||
More Information
Set Membership: |
|

Clp1
Genomic Map