Assay Details
| Gene Symbol: Casp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509419.2 | XP_003509467.1 | 4 - 5 | 563 | 72 | |
| XM_007650018.1 | XP_007648208.1 | 4 - 5 | 508 | 72 | ||
| GenBank mRNA | JI885300.1 | - | 1 - 2 | 109 | 72 | |
More Information
Set Membership: |
|

Casp1
Genomic Map