Assay Details
Gene Symbol: LOC100758426 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514977.2 | XP_003515025.2 | 3 - 4 | 1121 | 58 | |
XM_007654992.1 | XP_007653182.1 | 4 - 5 | 1002 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
