Assay Details
| Gene Symbol: Prune | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498658.2 | XP_003498706.1 | 2 - 3 | 848 | 96 | |
| XM_007631228.1 | XP_007629418.1 | 2 - 3 | 247 | 96 | ||
| XM_007631235.1 | XP_007629425.1 | 2 - 3 | 116 | 96 | ||
| GenBank mRNA | JI869040.1 | - | 2 - 3 | 224 | 96 | |
| JP042250.1 | - | 2 - 3 | 224 | 96 | ||

Prune
Genomic Map