Assay Details
Gene Symbol: Prune | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498658.2 | XP_003498706.1 | 4 - 5 | 1229 | 70 | |
XM_007631228.1 | XP_007629418.1 | 4 - 5 | 628 | 70 | ||
XM_007631235.1 | XP_007629425.1 | 4 - 5 | 497 | 70 | ||
GenBank mRNA | JI869040.1 | - | 4 - 5 | 605 | 70 | |
JP042250.1 | - | 4 - 5 | 605 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
