Assay Details
| Gene Symbol: Prune | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498658.2 | XP_003498706.1 | 6 - 7 | 1486 | 79 | |
| XM_007631228.1 | XP_007629418.1 | 6 - 7 | 885 | 79 | ||
| XM_007631235.1 | XP_007629425.1 | 6 - 7 | 754 | 79 | ||
| GenBank mRNA | JP042250.1 | - | 6 - 7 | 862 | 79 | |

Prune
Genomic Map