Assay Details
| Gene Symbol: Timm17a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498859.2 | XP_003498907.1 | 1 - 2 | 71 | 58 | |
| GenBank mRNA | JI890027.1 | - | 1 - 2 | 81 | 58 | |
| JP058933.1 | - | 1 - 2 | 81 | 58 | ||
More Information
Set Membership: |
|

Timm17a
Genomic Map