Assay Details
| Gene Symbol: Procr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501135.2 | XP_003501183.1 | 2 - 3 | 516 | 67 | |
| XM_007641834.1 | XP_007640024.1 | 1 - 2 | 247 | 67 | ||
| XM_007641835.1 | XP_007640025.1 | 2 - 3 | 525 | 67 | ||
| GenBank mRNA | JI869455.1 | - | 2 - 3 | 237 | 67 | |
| JP062235.1 | - | 2 - 3 | 237 | 67 | ||
More Information
Set Membership: |
|

Procr
Genomic Map