Assay Details
| Gene Symbol: Itfg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501559.2 | XP_003501607.1 | 12 - 13 | 1792 | 60 | |
| XM_007642122.1 | XP_007640312.1 | 12 - 13 | 1806 | 60 | ||
| GenBank mRNA | JI892196.1 | - | 3 - 4 | 334 | 60 | |
| JP054381.1 | - | 3 - 4 | 334 | 60 | ||
More Information
Set Membership: |
|

Itfg3
Genomic Map