Assay Details
| Gene Symbol: Itfg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501559.2 | XP_003501607.1 | 3 - 4 | 616 | 65 | |
| XM_007642122.1 | XP_007640312.1 | 3 - 4 | 630 | 65 | ||
| GenBank mRNA | JI878652.1 | - | 3 - 4 | 624 | 65 | |
| JP040363.1 | - | 3 - 4 | 624 | 65 | ||
More Information
Set Membership: |
|

Itfg3
Genomic Map