Assay Details
| Gene Symbol: Itfg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501559.2 | XP_003501607.1 | 5 - 6 | 925 | 66 | |
| XM_007642122.1 | XP_007640312.1 | 5 - 6 | 939 | 66 | ||
| GenBank mRNA | JI878652.1 | - | 5 - 6 | 933 | 66 | |
| JP040363.1 | - | 5 - 6 | 933 | 66 | ||
More Information
Set Membership: |
|

Itfg3
Genomic Map