Assay Details
| Gene Symbol: Itfg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501559.2 | XP_003501607.1 | 6 - 7 | 1061 | 71 | |
| XM_007642122.1 | XP_007640312.1 | 6 - 7 | 1075 | 71 | ||
More Information
Set Membership: |
|

Itfg3
Genomic Map